Learn More
Justrite™ SpillSlope™ Steel Shelf for 30 Gallon Safety Cabinet
Catalog No. 19822475 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 gal. Cabinet
Catalog No. For Use With (Equipment)
19-822-475 30 gal. Cabinet
1 options
Catalog No. 19-822-475 Supplier Justrite™ Supplier No. 29942
  • $231.00
    $157.65 / Each of 1
    Save $73.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 30 Gallon Safety Cabinet is a great way to add additional storage to facility cabinetry, expanding storage and spill protection.
Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump
Fits 30 gal. (36 in.W) cabinet
Specifications
For Use With (Equipment) 30 gal. Cabinet
Depth (English) Exterior 20 in.
Width (English) Exterior 32.375 in.
Material Steel