Learn More
Justrite™ SpillSlope™ Steel Shelf for 4 Gallon Safety Cabinet
GSA_VA
Catalog No. 19822476 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
4 gal. Cabinet
Catalog No. For Use With (Equipment)
19-822-476 4 gal. Cabinet
1 options
Catalog No. 19-822-476 Supplier Justrite™ Supplier No. 29935
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 4 Gallon Safety Cabinet is a great way to add additional storage to facility cabinetry, expanding storage and spill protection.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Fits 4 gal. cabinets
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) 4 gal. Cabinet
Material Steel
Depth (English) Exterior 13 in.
Width (English) Exterior 13.375 in.