Learn More
Justrite™ SpillSlope™ Steel Shelf for 54 Gallon Deep Slimline Safety Cabinet
Catalog No. 19822482 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
54 gal. Cabinet
Catalog No. For Use With (Equipment)
19-822-482 54 gal. Cabinet
1 options
Catalog No. 19-822-482 Supplier Justrite™ Supplier No. 29941
  • $234.50
    $168.65 / Each of 1
    Save $65.85
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 54 Gallon Deep Slimline Safety Cabinet adds additional storage to cabinets.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Fits 54 gal. Deep Slimline cabinet
Specifications
For Use With (Equipment) 54 gal. Cabinet
Depth (English) Exterior 29 in.
Material Steel
Width (English) Exterior 19.625 in.