Learn More
Justrite™ SpillSlope™ Steel Shelf for 90 Gallon Safety Cabinet
Catalog No. 19822477 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
90 gal. Cabinet
Catalog No. For Use With (Equipment)
19-822-477 90 gal. Cabinet
1 options
Catalog No. 19-822-477 Supplier Justrite™ Supplier No. 29945
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 90 Gallon Safety Cabinet is a great way to add additional storage to facility cabinetry, expanding storage and spill protection.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Fits 90 gal. (43 in.W) cabinet
Specifications
For Use With (Equipment) 90 gal. Cabinet
Depth (English) Exterior 29 in.
Width (English) Exterior 39.375 in.
Material Steel