Learn More
Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet
Catalog No. 19822484 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
15 gal. Under Fume Hood Cabinet
19 gal. Under Fume Hood Cabinet
23 gal. Under Fume Hood Cabinet
31 gal. Under Fume Hood Cabinet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-822-484 15 gal. Under Fume Hood Cabinet
19-822-485 19 gal. Under Fume Hood Cabinet
19-822-486 23 gal. Under Fume Hood Cabinet
19-822-487 31 gal. Under Fume Hood Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-822-484 Supplier Justrite™ Supplier No. 29950
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet adds additional storage to cabinets.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Fits 15-gallon Under Fume Hood cabinets.
  • Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs. Galvanized steel shelves hold up to 350 lb. (159kg) and adjust on 3 in. (76-mm) increments for versatile storage.
  • Meets ANSI MH28.1
Specifications
For Use With (Equipment) 15 gal. Under Fume Hood Cabinet