Learn More
Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet
Catalog No. 19822487 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
15 gal. Under Fume Hood Cabinet
19 gal. Under Fume Hood Cabinet
23 gal. Under Fume Hood Cabinet
31 gal. Under Fume Hood Cabinet
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-822-487 31 gal. Under Fume Hood Cabinet
19-822-484 15 gal. Under Fume Hood Cabinet
19-822-485 19 gal. Under Fume Hood Cabinet
19-822-486 23 gal. Under Fume Hood Cabinet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-822-487 Supplier Justrite™ Supplier No. 29953
  • $278.00
    $187.65 / Each of 1
    Save $90.35
    33% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf For Under Fume Hood Safety Cabinet adds additional storage to cabinets.
  • Protect workers, Reduce fire risks, and improve productivity by storing flammable fuels and chemicals in code-compliant safety cabinets
  • Designed to meet OSHA and NFPA standards, Sure-Grip EX cabinets are constructed of sturdy 18-gauge thick double-wall welded steel with 1-1/2 in. of insulating air space for fire resistance
  • High performance, self-latching doors close easily and securely for maximum protection under fire conditions
  • Fail-safe closing mechanism ensures three-point stainless steel bullet latching system works every time
  • For added security, U-Loc handle comes with a cylinder lock and key set or accepts an optional padlock
  • Haz-Alert reflective warning labels are highly visible under fire conditions or during power outages when illuminated with a flashlight
  • Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof bottom sump
  • They adjust on 3 in. centers for versatile storage
  • Cabinets also feature dual vents with flame arresters, four adjustable self-leveling feet, grounding connector, and trilingual warning label
  • Durable lead-free epoxy/polyester powder-coat finish provides excellent chemical resistance
  • The ChemCor thermoplastic coating on all interior surfaces provides superior protection against corrosion from drips and off-gassing from harsh chemicals
  • Seam-free surface eliminates the need for rivets or fasteners which risk corrosion from seep-through chemicals
  • Non-porous surface resists staining and cleans up easily
Specifications
For Use With (Equipment) 31 gal. Under Fume Hood Cabinet