Learn More
Justrite™ SpillSlope™ Steel Shelf with Poly Tray for 12/15 Gallon Compac and 22 Gallon Slimline Safety Cabinets
Catalog No. 19822504 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
Fits 12 & 15 gal. Compac Cabinet and 22 gal. Slimline Cabinet
Catalog No. For Use With (Equipment)
19-822-504 Fits 12 & 15 gal. Compac Cabinet and 22 gal. Slimline Cabinet
1 options
Catalog No. 19-822-504 Supplier Justrite™ Supplier No. 29959
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf with Poly Tray for 12/15 Gallon Compac and 22 Gallon Slimline Safety Cabinets is a great combo that provides additional storage and spill protection.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • These Polyethylene accessories with slightly curved edge provide additional protection against leaks and spills
Specifications
Dimensions (D x W x H) 14 x 19.625 in.
For Use With (Equipment) Fits 12 & 15 gal. Compac Cabinet and 22 gal. Slimline Cabinet
Material Steel, Polyethylene