Learn More
Justrite™ SpillSlope™ Steel Shelf with Polyethylene Tray for 17/30/45 Gallon Safety Cabinets
Shop All Justrite Safety Group ProductsDescription
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
- These Polyethylene accessories with slightly curved edge provide additional protection against leaks and spills
Specifications
Specifications
| For Use With (Equipment) | 17/30/45 gal. Cabinets |
| Depth (English) Exterior | 14 in. |
| Material | Steel, Polyethylene |
| Width (English) Exterior | 39.375 in. |