Learn More
Justrite™ SpillSlope™ Steel Shelf with Polyethylene Tray for 22 Gallon Undercounter Safety Cabinet
Catalog No. 19822490 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
22 gal. Undercounter Cabinet
Catalog No. For Use With (Equipment)
19-822-490 22 gal. Undercounter Cabinet
1 options
Catalog No. 19-822-490 Supplier Justrite™ Supplier No. 29960
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf with Polyethylene Tray for 22 Gallon Undercounter Safety Cabinet is the perfect product to add additional storage space. The Chemchor™ thermoplastic liner offers superior protection.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • These Polyethylene accessories with slightly curved edge provide additional protection against leaks and spills
Specifications
For Use With (Equipment) 22 gal. Undercounter Cabinet
Width (English) Exterior 31.625 in.
Material Steel, Polyethylene
Depth (English) Exterior 18 in.