Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8
SDP
Catalog No. CG200103
Encompass_Preferred
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8. Octagon cross-section having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.SEE CG-2001-50 FOR STIR BAR KIT
Catalog No. CG200103 Supplier Chemglass Life Sciences Supplier No. CG200103
Stir Bar, Magnetic, PTFE, Length x Dia. in mm: 12.7 x 9.52, Length x Dia. in inches: 1/2 x 3/8. Octagon cross-section having a molded-in pivot ring manufactured with FDA-grade and USP Class VI PTFE. The PTFE coated Alnico magnet permits magnetic agitation without contamination.SEE CG-2001-50 FOR STIR BAR KIT