Learn More
Electron Microscopy Sciences 13 Egg-Shaped Spin Bar Assortment Bx
SDP
Catalog No. 501899178
Encompass_Preferred
Egg-shaped magnetic stirring bars provide own pivot point. Designed for use in round-bottom vessels, 13 Egg-shaped spin bars, PTFE, consists 1 each: 3" x 3/4", 2 each: 3/4 "x 3/8", 1 "x 1/2", 1/4" x 5/8", 1.5" x 5/8 ", 1 5/8 x 3/4", 2" x 3/4".
Catalog No. 50-189-9178 Supplier Electron Microscopy Sciences Supplier No. 6451013
Egg-shaped magnetic stirring bars provide own pivot point. Designed for use in round-bottom vessels, 13 Egg-shaped spin bars, PTFE, consists 1 each: 3" x 3/4", 2 each: 3/4 "x 3/8", 1 "x 1/2", 1/4" x 5/8", 1.5" x 5/8 ", 1 5/8 x 3/4", 2" x 3/4".