Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Egg-shaped, fits 3000mL-5000mL flask, Length x Dia. in inches: 2 1/2 x 3/4
SDP
Catalog No. CG200516
Encompass_Preferred
Stir Bar, Magnetic, Egg-shaped, fits 3000mL-5000mL flask, Length x Dia. in inches: 2 1/2 x 3/4. Egg-shaped magnetic stirring bar provides its own pivot point without separate ring or abrupt change in contour. Designed for use in round bottom vessels, it is shaped to spin freely, unrestricted by the vessel contour. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2005-20 FOR STIR BAR KIT
Catalog No. CG200516 Supplier Chemglass Life Sciences Supplier No. CG200516
Stir Bar, Magnetic, Egg-shaped, fits 3000mL-5000mL flask, Length x Dia. in inches: 2 1/2 x 3/4. Egg-shaped magnetic stirring bar provides its own pivot point without separate ring or abrupt change in contour. Designed for use in round bottom vessels, it is shaped to spin freely, unrestricted by the vessel contour. Manufactured with FDA-grade and USP Class VI PTFE.SEE CG-2005-20 FOR STIR BAR KIT