Learn More
Chemglass Life Sciences Stir Bar, Magnetic, Glass, Length x Dia. in mm: 38.1 x 9.52, Length x Dia. in inches: 1-1/2 x 3/8
SDP
Catalog No. 50872473
Encompass_Preferred
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 38.1 x 9.52, Length x Dia. in inches: 1-1/2 x 3/8. Borosilicate covered Alnico magnet for use in high temperature reactions.
Catalog No. 50-872-473 Supplier Chemglass Life Sciences Supplier No. CG200413
Stir Bar, Magnetic, Glass, Length x Dia. in mm: 38.1 x 9.52, Length x Dia. in inches: 1-1/2 x 3/8. Borosilicate covered Alnico magnet for use in high temperature reactions.