Learn More
Electron Microscopy Sciences 16 Spin Bars Assortment Box
SDP
Catalog No. 501899179
Encompass_Preferred
Box contains 16 assorted bars; 12 octagon magnetic stirring bars, PTFE with pivot ring and 4 fleas, 2 each: 7mmx2mm, 10mmx3mm, 1/2"x 5/16", 5/8 "x 5/16", 1"x3/8", 1.5"x 3/8", 2"x 3/8", 2.5"x 3/8".
Catalog No. 50-189-9179 Supplier Electron Microscopy Sciences Supplier No. 6451016
Box contains 16 assorted bars; 12 octagon magnetic stirring bars, PTFE with pivot ring and 4 fleas, 2 each: 7mmx2mm, 10mmx3mm, 1/2"x 5/16", 5/8 "x 5/16", 1"x3/8", 1.5"x 3/8", 2"x 3/8", 2.5"x 3/8".