Learn More
Boca Scientific Inc MobiSpin Column with screw cap, inserted large 10μm filter and bottom plug, 100/pack
Supplier:  Boca Scientific Inc M1002S
Encompass
The M1002S MobiSpin Column is an empty spin column with a 10 µm pore-size large filter, a screw cap and an inserted bottom plug. These Mobicols can be used to make your own spin/affinity columns with the matrix of your choice. As Spin Columns, M1002S Mobicols are placed in a 1.5ml microtube and can be centrifuged in a microcentrifuge. These columns are easy to handle and can treat both small and large volumes.
Catalog No. NC1472174