Learn More
Alkali Scientific Spin Mini Prep High purity Kit
SDP
Supplier:  Alkali Scientific C605
Encompass_Preferred
Spin Mini Prep High Purity Kit is designed for fast and efficient DNA purification from plasmid samples. This kit includes high quality Spin Columns, Reagents, Buffers, and Collection Tubes necessary for optimal nucleic acid extraction. The Spin Columns utilize a silica based membrane to ensure high binding capacity and purity during DNA isolation. It efficiently isolates DNA from contaminants like proteins, salts, and other impurities. The buffer system ensures effective removal of contaminants, providing high yields of clean, intact DNA. This kit is perfect for small scale DNA purifications and is suitable for cloning, sequencing, and molecular biology applications. It includes easy to follow instructions and comes in a 100 pack for high volume labs. The reagents and buffers included ensure consistent, reliable performance in every prep. The kit is ideal for quick and accurate plasmid DNA purification.
Catalog No. 50-006-62614