Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Spin Tip

Catalog No. 89339127 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Equipment):
SpiralPipet
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
89-339-127 SpiralPipet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-127 Supplier Abnova™ Supplier No. U0384
Add to Cart
Add to Cart

Spin Tip is used for applications with SpiralPipet™ instrument.

Specifications

Storage Requirements Store at room temperature.
Quantity 1000 Slides
For Use With (Equipment) SpiralPipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.