Learn More
Chemglass Life Sciences Stir Bar, Magnetic, PTFE, "V" Shaped, 16mm Height x 10mm Diameter
SDP
Catalog No. CG200811
Encompass_Preferred
Stir Bar, Magnetic, PTFE, "V" Shaped, 16mm Height x 10mm Diameter. "V" Shaped PTFE magnetic stir bar. Design permits rotation in small test tubes and conical vessels such as MW-80 reaction vials. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. CG200811 Supplier Chemglass Life Sciences Supplier No. CG200811
Stir Bar, Magnetic, PTFE, "V" Shaped, 16mm Height x 10mm Diameter. "V" Shaped PTFE magnetic stir bar. Design permits rotation in small test tubes and conical vessels such as MW-80 reaction vials. Manufactured with FDA-grade and USP Class VI PTFE.