Learn More
Sigma Aldrich Fine Chemicals Biosciences Corning(R) Costar(R) Spin-X(R) centrifuge tube filters Nylon membrane, pore size 0.45 mum, non-sterile | 200EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS8170200EA
Encompass
Corning(R) Costar(R) Spin-X(R) centrifuge tube filters Nylon membrane, pore size 0.45 mum, non-sterile | 200EA
Catalog No. NC2165959