Learn More
Aviva Systems Biology Corp SPIN1 Antibody - middle region (ARP52248_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52248_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPIN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SPIN1. Alternate names: SPINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-672