Learn More
Electron Microscopy Sciences Tissue Matrice Spinal Cord Coronal & Sagittal, 1 mm Mouse, 4 mm W x 50 mm L Cavity, 5 mm Depth
SDP
Catalog No. 501928517
Encompass_Preferred
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 1 mm Mouse
Cavity: 4 mm W x 50 mm L
Depth: 5 mm
Catalog No. 50-192-8517 Supplier Electron Microscopy Sciences Supplier No. 69085CS
These spinal cord matrices are precisely machined to ensure reproducible 0.5mm or 1mm sections of mouse or rat spinal cords.
Type: 1 mm Mouse
Cavity: 4 mm W x 50 mm L
Depth: 5 mm