Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinal Diluting Fluid, Crystal Violet-Acetic Acid Solution for Cerebrospinal Fluid Cell Count, Ricca Chemical
SDP

Catalog No. 79964 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7996-4 120 mL Natural Poly Bottle
7996-16 500 mL Natural Poly Bottle
799632 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7996-4 Supplier Ricca Chemical Company Supplier No. 79964
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Physical Form Solution
Chemical Name or Material SPINAL DILUTING FLUID Crystal Violet- Acetic Acid Solution
CAS 7732-18-5, 64-19-7, 548-62-9
CAS Min % 90.0, 7.88, 0.1
CAS Max % 94.0, 8.2, 0.11
Packaging Natural Poly Bottle
Color Dark Violet
pH <1
Quantity 120 mL
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.