Learn More
Medchemexpress LLC HY-N6962 1mg Medchemexpress, α-Spinasterol CAS:481-18-5 Purity:>98%
SDP
Catalog No. 502082976
Encompass_Preferred
Medchemexpress, HY-N6962 1mg α-Spinasterol CAS:481-18-5 α-Spinasterol, isolated from Spinacia oleracea, has antibacterial activity. α-Spinasterol is a transient receptor potential vanilloid 1 (TRPV1) antagonist, has anti-inflammatory, antidepressant, antioxidant and antinociceptive effects. α-Spinasterol inhibits COX-1 andCOX-2 activities with IC50 values of 16.17 μM and 7.76 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2976 Supplier Medchemexpress LLC Supplier No. HYN69621MG
Medchemexpress, HY-N6962 1mg α-Spinasterol CAS:481-18-5 α-Spinasterol, isolated from Spinacia oleracea, has antibacterial activity. α-Spinasterol is a transient receptor potential vanilloid 1 (TRPV1) antagonist, has anti-inflammatory, antidepressant, antioxidant and antinociceptive effects. α-Spinasterol inhibits COX-1 andCOX-2 activities with IC50 values of 16.17 μM and 7.76 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.