Learn More
Medchemexpress LLC α-Spinasterol | 481-18-5 | MFCD00200554 | 5 MG
SDP
Catalog No. 5000417985
Encompass_Preferred
α-Spinasterol is an orally taken antagonist of transient receptor potential vanilloid 1 (TRPV1), and it is also an inhibitor of COX-1 and COX-2, with IC50 values of 16.17 μM and 7.76 μM, respectively. It exhibits antibacterial, anti-inflammatory, antidepressant, and antioxidant effects, has the ability to cross the blood-brain barrier, and can improve diabetes in mice.
  • Antagonist of transient receptor potential vanilloid 1 (TRPV1)
  • Inhibitor of COX-1 and COX-2
  • Exhibits antibacterial, anti-inflammatory, antidepressant, and antioxidant effects
  • Ability to cross the blood-brain barrier
  • Can improve diabetes in mice
Catalog No. 50-004-17985 Supplier Medchemexpress LLC Supplier No. HYN69625MG
α-Spinasterol is an orally taken antagonist of transient receptor potential vanilloid 1 (TRPV1), and it is also an inhibitor of COX-1 and COX-2, with IC50 values of 16.17 μM and 7.76 μM, respectively. It exhibits antibacterial, anti-inflammatory, antidepressant, and antioxidant effects, has the ability to cross the blood-brain barrier, and can improve diabetes in mice.
  • Antagonist of transient receptor potential vanilloid 1 (TRPV1)
  • Inhibitor of COX-1 and COX-2
  • Exhibits antibacterial, anti-inflammatory, antidepressant, and antioxidant effects
  • Ability to cross the blood-brain barrier
  • Can improve diabetes in mice