Learn More
Andwin Scientific SPINBAR DISP 1/2D X 1/2L
SDP
Catalog No. NC0580151
Encompass_Preferred
The Spinbar are disposable, multifaceted stir bars that increase turbulence relative to similar size smooth cylindrical stirring bars. This product is recommended for use in single use applications, such as mixing radioactive reagents. The stirring
Catalog No. NC0580151 Supplier Andwin Scientific Supplier No. 58947140
The Spinbar are disposable, multifaceted stir bars that increase turbulence relative to similar size smooth cylindrical stirring bars. This product is recommended for use in single use applications, such as mixing radioactive reagents. The stirring