Learn More
Research Products International Corp Spinbar Magnetic Sink Strainer, Blue
SDP
Catalog No. 501978359
Encompass
The Spinbar Magnetic Sink Strainer provides an innovative solution to a common laboratory inconvenience. Prevents magnetic stirring bars from accidentally slipping into the drain when cleaning beakers or cylinders. Flexible chemical resistant strainer has four strong magnets that attract and hold stirring bars. The strainer is designed with long slits that allow unobstructed drainage of fluids. Strainer remains securely in place and fits a variety of sinks. Flat edge allows strainer to be placed near the sink wall.
Catalog No. 50-197-8359 Supplier Research Products International Corp Supplier No. 249013
The Spinbar Magnetic Sink Strainer provides an innovative solution to a common laboratory inconvenience. Prevents magnetic stirring bars from accidentally slipping into the drain when cleaning beakers or cylinders. Flexible chemical resistant strainer has four strong magnets that attract and hold stirring bars. The strainer is designed with long slits that allow unobstructed drainage of fluids. Strainer remains securely in place and fits a variety of sinks. Flat edge allows strainer to be placed near the sink wall.