Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR disposable magnet
Catalog No. 502900805
Encompass
for single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Catalog No. 50-290-0805 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4202201PAK
for single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.