Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR disposable magnet
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z4202391PAK
Encompass
for single-use application where efficiency and cross-contamination are a concern. Alnico V magnet.
Catalog No. 50-290-0828