Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR magnetic stir bar1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1270191EA
Encompass
Leakproof seamless polished and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Catalog No. 50-290-1546