Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR magnetic stir bar1EA
Catalog No. 502900963
Encompass
Leakproof seamless polished and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
Catalog No. 50-290-0963 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1044421EA
Leakproof seamless polished and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.