Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR magnetic stirring
Catalog No. 502900062
Encompass
Very small efficient stirring bars. Ideal for vials micro test tubes centrifuge tube mixing etc.PTfE(R) PTfE coatedALNICO V coated magnet
Catalog No. 50-290-0062 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2838273EA
Very small efficient stirring bars. Ideal for vials micro test tubes centrifuge tube mixing etc.PTfE(R) PTfE coatedALNICO V coated magnet