Learn More
Sigma Aldrich Fine Chemicals Biosciences SpinbarR magnetic stirring
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2838193EA
Encompass
Very small efficient stirring bars. Ideal for vials micro test tubes centrifuge tube mixing etc.PTfE(R) PTfE coatedALNICO V coated magnet
Catalog No. 50-290-1381