Learn More
Anton Paar Medium-viscosity Spindle Guard R for ViscoQC 100-R and ViscoQC 300-R
Catalog No. NC1818596
Encompass_Preferred
Spindle guard made of stainless steel AISI 316L (1.4404 / 1.4435). Comes with a fixing screw with identification """"R"""". The screw also serves as guard detection, which avoids measurement with a wrong configuration (TruGuard). For use inside wide-rimmed vessels (typically 600 mL glass beakers). Required for spindles RH1 and RH2.Measurement Spindle RH1 item NC1818591Measurement Spindle RH2 included with ViscoQCSuitable for use with Anton Paar rotational viscometers ViscoQC 100 – R item NC1818567, ViscoQC 300 – R item NC1818576.
Catalog No. NC1818596 Supplier Anton Paar Supplier No. 191718
Spindle guard made of stainless steel AISI 316L (1.4404 / 1.4435). Comes with a fixing screw with identification """"R"""". The screw also serves as guard detection, which avoids measurement with a wrong configuration (TruGuard). For use inside wide-rimmed vessels (typically 600 mL glass beakers). Required for spindles RH1 and RH2.Measurement Spindle RH1 item NC1818591Measurement Spindle RH2 included with ViscoQCSuitable for use with Anton Paar rotational viscometers ViscoQC 100 – R item NC1818567, ViscoQC 300 – R item NC1818576.