Learn More
Brookfield Engineer Labs LV-5 Standard Spindles
Catalog No. NC1250422
Encompass
Standard LV spindles to be used with your Brookfield Viscometer or Rheometer with an LV torque range.
Catalog No. NC1250422 Supplier Brookfield Engineer Labs Supplier No. LV5
Standard LV spindles to be used with your Brookfield Viscometer or Rheometer with an LV torque range.