Learn More
Anton Paar Spindle Rack for ViscoQC Spindles
Catalog No. NC1818599
Encompass_Preferred
A rack made of stainless steel with 7 positions. Prevents damage to spindles, which are not in use. Suitable for any spindle type. Equipped with anti-slip rubber feet.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818599 Supplier Anton Paar Supplier No. 189441
A rack made of stainless steel with 7 positions. Prevents damage to spindles, which are not in use. Suitable for any spindle type. Equipped with anti-slip rubber feet.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.