Learn More
Anton Paar Measuring spindle V74 - Low volume vane spindle for ViscoQC 100 & ViscoQC 300
Catalog No. NC1818341
Encompass_Preferred
Stainless steel vane spindle equipped with Toolmaster and magnetic coupling. Intended for measuring gel like or shear sensitive substances. Full immersion only for very low sample volume in 25 ml beaker. Recommended 20 ml sample volume.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818341 Supplier Anton Paar Supplier No. 198723
Stainless steel vane spindle equipped with Toolmaster and magnetic coupling. Intended for measuring gel like or shear sensitive substances. Full immersion only for very low sample volume in 25 ml beaker. Recommended 20 ml sample volume.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.