Learn More
Brookfield Engineer Labs SPINDLE,VISCOMETER,RV-5,302SS
Catalog No. NC1581008
Encompass
SPINDLE, VISCOMETER, RV-5, 302 SS, SERIES RANGE RV 400-800K, HA 800-1.6M, HB 3.2K-6.4M
Catalog No. NC1581008 Supplier Brookfield Engineer Labs Supplier No. RVHV5
SPINDLE, VISCOMETER, RV-5, 302 SS, SERIES RANGE RV 400-800K, HA 800-1.6M, HB 3.2K-6.4M