Learn More
Anton Paar Set of vane spindles for ViscoQC 100 & ViscoQC 300
Catalog No. NC1818340
Encompass_Preferred
Set of 3 stainless steel vane spindles V71, V72, V73, equipped with Toolmaster and magnetic coupling. Intended for measuring gel like or shear sensitive substances. Come with immersion marks for full or half immersion depth.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818340 Supplier Anton Paar Supplier No. 198919
Set of 3 stainless steel vane spindles V71, V72, V73, equipped with Toolmaster and magnetic coupling. Intended for measuring gel like or shear sensitive substances. Come with immersion marks for full or half immersion depth.Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.