Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ 96-Well DNA Kit for Blood

Catalog No. MP116552096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
4 x 96 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116552096 96 Preps
MP116552496 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116552096 Supplier MP Biomedicals Supplier No. 116552096
Add to Cart
Add to Cart

Designed for high-throughput genomic DNA extraction, processing 96 samples from blood simultaneously.

  • Compatible with fresh and frozen human whole blood preserved in EDTA, sodium citrate, or heparin
  • Provides high-yield, high-purity gDNA ideal for long-fragment PCR, real-time PCR, restriction enzyme digestion, and sequencing
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Column
Scale Mini
No. of Reactions 96 Preps
Product Type DNA Kit for Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.