Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ 96-Well DNA Kit for Tissue

Catalog No. MP116559096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
4 x 96 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116559096 96 Preps
MP116559496 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116559096 Supplier MP Biomedicals Supplier No. 116559096
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to efficiently isolate high-quality genomic DNA from a wide range of tissue types using a streamlined workflow.

The SPINeasy™ 96-Well DNA Kit for Tissue is a high-performance tissue genomic DNA (gDNA) extraction kit in a 96-well plate format. The DNA purification protocol does not require any organic solvent or sophisticated equipment. Our specially formulated Buffer TD1 and Proteinase K provide effective lysis of various types of samples including tough, fibrous, and fatty tissues. Furthermore, the subsequent addition of Buffer TD2 enables selective binding of DNA to 96 DNA Plate M. The gDNA extracted from multiple types of tissues using this kit showed optimal integrity and purity which can immediately be used for most downstream applications, including long fragment PCR, real time PCR, restriction enzyme digestion and sequencing.

  • Enables high-yield extraction of genomic DNA from multiple tissue types, including tough, fibrous, and fatty samples
  • Produces high-molecular-weight genomic DNA greater than 50 kb with excellent purity
  • Purified DNA is suitable for a wide range of downstream applications, including long-fragment PCR, real-time PCR, restriction digestion, and sequencing
  • High-capacity 96-well DNA binding plate supports consistent performance and parallel sample processing
  • Optimized lysis chemistry using proprietary buffers and Proteinase K ensures effective tissue disruption and DNA release
  • Streamlined protocol requires no organic solvents or specialized equipment and delivers results with less than 40 minutes of hands-on time
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 96 Preps
Product Type DNA Kit for Tissue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.