Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ 96-Well DNA Kit for Tissue

Catalog No. MP116559496 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
4 x 96 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116559496 4 x 96 Preps
MP116559096 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116559496 Supplier MP Biomedicals Supplier No. 116559496
Only null left
Add to Cart
Add to Cart

Designed to efficiently isolate high-quality genomic DNA from a wide range of tissue types using a streamlined workflow.

  • Enables high-yield extraction of genomic DNA from multiple tissue types, including tough, fibrous, and fatty samples
  • Produces high-molecular-weight genomic DNA greater than 50 kb with excellent purity
  • Purified DNA is suitable for a wide range of downstream applications, including long-fragment PCR, real-time PCR, restriction digestion, and sequencing
  • High-capacity 96-well DNA binding plate supports consistent performance and parallel sample processing
  • Optimized lysis chemistry using proprietary buffers and Proteinase K ensures effective tissue disruption and DNA release
  • Streamlined protocol requires no organic solvents or specialized equipment and delivers results with less than 40 minutes of hands-on time
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 4 x 96 Preps
Product Type DNA Kit for Tissue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.