Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ 96-Well Plasmid Miniprep Kit

Catalog No. MP116534096 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
4 x 96 Preps
96 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116534096 96 Preps
MP116534496 4 x 96 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP116534096 Supplier MP Biomedicals Supplier No. 116534096
Add to Cart
Add to Cart

High-throughput purification method for recovering plasmid DNA in less than 45 minutes.

  • Effective binding method capable of accommodating up to 4 mL of bacterial culture, yielding up to 100 μg of plasmid DNA.
  • Fully compatible with downstream applications such as restriction enzyme digestion and endpoint PCR.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Column
Scale Mini
No. of Reactions 96 Preps
Product Type Plasmid Miniprep Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.