Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Blood

Catalog No. MP116552050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116552050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116552050 Supplier MP Biomedicals™ Supplier No. 116552050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fast and efficient DNA isolation from blood and other biofluids in less than 30 minutes. Convenient spin-column technology with no organic solvents. High DNA yield and purity, free of common contaminants. DNA ready for downstream PCR or NGS applications.

  • SPINeasy DNA Kit for Blood is a high-performance genomic DNA (gDNA) extraction kit based on silica-membrane spin-column technology.
  • This kit enables quick isolation of gDNA from whole blood preserved in different anticoagulants (EDTA, heparin, and sodium citrate), typically in less than 30 minutes.
  • It also allows easy gDNA isolation from coagulated blood when using the FastPrep-24 5G homogenizer with lysing matrix A or D tubes (recommended for soft tissues, and not included with this kit).
  • The use of our specially formulated Lysis Buffer BL enables highly efficient lysis of various liquid samples including plasma, serum, saliva, and cell culture medium.
  • Column S1 and kit buffers (provided in the kit) are designed to deliver gDNA extracts of high yield and purity, compatible with downstream molecular applications such as PCR and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Includes Lysis Buffer BL, Wash Buffer BW1, Wash Buffer BW2, Elution Buffer EB, Column S, 2.0 mL collection tubes,1.5 mL microcentrifuge tubes, Proteinase K, Proteinase K Buffer
Product Type DNA Extraction Kit
Quantity 50 Preps
Sample Volume 200 µL of Human Blood, Serum, Plasma, Saliva, or Cell Culture Media

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.