Learn More
Description
- SPINeasy DNA Kit for Microbiome allows for efficient and unbiased lysis of microbes, including gram positive and negative bacteria, fungi, protozoans, and viruses found in various swab, body fluid, stool, soil, and water samples.
- This kit features a highly effective lysis system and silica membrane spin column technology. With the use of specially formulated Buffer MB1 and Lysing Matrix E in combination with FastPrep instruments from MP Biomedicals, efficient lysis of various samples can be achieved within seconds.
- Provided with the kit, Column MB and kit buffers are designed to deliver gDNA of high yield and purity in fewer steps, ready for downstream applications such as qPCR, restriction digestion, and sequencing.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Dry Environment |
| Format | Kit |
| Isolation Technology | Spin Column |
| Sample Type | Swabs, Sputum, Body Fluids, Stool, and Environmental Samples (e.g. soil and water) |
| Includes | Lysing Matrix E (50 ea), RNase A Solution (2 mL), Buffer MB1 (45 mL), Buffer MB2 (17 mL), Buffer MB3 (21 mL × 2), Buffer MB4 (28 mL), Buffer MB5 (20 mL × 2), Buffer MB6 (10 mL), Column MB (50 ea), 1.5 mL Collection Tubes (50 ea) |
| Product Type | DNA Extraction Kit |
| Quantity | 50 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.