Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Tissue and Bacteria

Catalog No. MP116532050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116532050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116532050 Supplier MP Biomedicals™ Supplier No. 116532050

missing translation for 'validMainframeMsg'

Fast and easy isolation of high-quality genomic DNA from animal tissues, blood, cultured cells, and bacteria. Lysing Matrix A for thorough and complete lysis with FastPrep Instruments. High-performance gDNA extraction kit based on silica membrane spin column technology.

  • SPINeasy DNA Kit for Tissue and Bacteria is a high-performance genomic DNA (gDNA) extraction kit based on silica membrane spin column technology.
  • The kit enables quick isolation of gDNA from various animal tissues, blood, cultured cells, and bacteria, typically in less than 30 minutes.
  • The use of our specially formulated Lysis Buffer GD and Lysing Matrix A in combination with FastPrep instruments from MP Biomedicals enables highly efficient lysis of various tissue types within seconds.
  • The provided Column GD coupled with specially formulated kit buffers are designed to deliver gDNA of high yield and purity with fewer steps, suitable for downstream applications such as restriction digestion, PCR, and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type Blood, Tissue, Bacteria, Insects, Swabs
Includes 50 x 2 mL Lysing Matrix A, 60 mL Lysis Buffer GD, 30 mL Wash Buffer GD1, 6 mL Wash Buffer GD2, 10 mL Elution Buffer GD, 50 x Column GD with collection tubes
Product Type DNA Extraction Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.