Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ DNA Kit for Tissue (with Lysing Matrix)

Catalog No. MP116558050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116558050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116558050 Supplier MP Biomedicals Supplier No. 116558050
Add to Cart
Add to Cart

High-performance genomic DNA extraction kit combining mechanical, chemical, and enzymatic tissue disruption in 30 minutes.

  • The addition of proteinase K allows a more efficient and complete digestion of any tissue type.
  • Homogenizing time is greatly reduced while substantially increasing the gDNA yield and purity.
  • The kit is validated for tissue samples of fatty, tough, and fibrous samples, and suitable for downstream applications such as PCR and qPCR.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 50 Preps
Product Type DNA Kit for Tissue (with Lysing Matrix)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.