Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ DNA Kit for Urine

Catalog No. MP116593050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116593050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116593050 Supplier MP Biomedicals Supplier No. 116593050
Add to Cart
Add to Cart

Allows simultaneous isolation of both genomic DNA (gDNA) and cell-free DNA (cfDNA) from a single urine sample

  • Enables 30-day ambient storage when used with the Urine Collection and Preservation Kit (Cat. No. 116594000), yielding high-quality DNA suitable for PCR, qPCR, and sequencing Minimal handling enables fast and reliable results in any lab setting
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 50 Preps
Product Type DNA Kit for Urine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.