Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA Kit for Water

Catalog No. MP116536050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116536050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116536050 Supplier MP Biomedicals™ Supplier No. 116536050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifically designed for the isolation of microbial DNA from various water samples. Fast and easy extraction of gDNA in less than 30 minutes. Supplied with filter membranes and Lysing matrix tubes to capture and concentrate microbes and efficiently homogenize in using a FastPrep instrument.

  • SPINeasy DNA Kit for Water is a high-performance water genomic DNA extraction kit based on silica membrane spin column technology.
  • The kit has been specifically designed to concentrate microbes present in water samples by filtration and transfer to a Lysing Matrix tube (4.5 mL) for the best extraction of microbial gDNA from water in less than 30 min.
  • Water samples are processed using our uniquely formulated Lysis Buffer W1 and Lysing Matrix E to effectively lyse various types of microbial cells, including bacteria, fungi, virus, and protozoans.
  • Column W1 (provided with the kit) has high binding capacity and selectivity for gDNA.
  • The combination of components in the kit helps extract microbial gDNA of high yield and purity that is ready for downstream applications, such as restriction digestion, PCR, and NGS.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type Water Samples
Includes 50 x 5 mL Lysing Matrix E, 60 mL Lysis Buffer W1, 8 mL Lysis Buffer W2, 550 µL RNase A Solution, 15 mL Inhibitor Removal W, 30 mL Binding Buffer W, 9 mL Wash Buffer W1, 6 mL Wash Buffer W2, 10 mL DES Buffer, 50 x Filter Membrane, 50 x Column W1, 50 x 2.0 mL Collection Tubes, 50 x 1.5 mL Collection Tubes
Product Type DNA Isolation Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.