Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals SPINeasy™ DNA Kit for Yeast

Catalog No. MP116557050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116557050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116557050 Supplier MP Biomedicals Supplier No. 116557050
Add to Cart
Add to Cart

This kit enables quick isolation of gDNA from yeast cells, typically in less than 30 minutes.

  • High-performance genomic DNA (gDNA) extraction kit which is developed based on silica-membrane spin-column technology.
TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
Isolation Technology Silica Spin Column
Scale Mini
No. of Reactions 50 Preps
Product Type DNA Kit for Yeast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.