Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ SPINeasy™ DNA Pro Kit for Feces

Catalog No. MP116547050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116547050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116547050 Supplier MP Biomedicals™ Supplier No. 116547050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Efficient and rapid isolation of high-quality genomic DNA from both fecal and soil samples. Optimized Buffer SF3 enables specific binding of DNA without RNA contamination. New Lysing Matrix YB tubes and Lysis Buffers provide efficient and reproducible sample lysis with FastPrep instruments.

  • The SPINeasy DNA Pro Kit for Feces efficiently isolates high-quality gDNA from stool samples. Fecal samples present distinct sample processing challenges due to sample composition which is largely dependent on diet and may include fibers, undigested particles, bilirubin, complex polysaccharides, and lipids. These compounds may impair sample homogenization, decreasing both the quantity and quality of the extracted DNA.
  • The SPINeasy DNA Pro Kit for Feces was designed to address these challenges. Samples are optimally homogenized by bead beating with new Lysing Matrix YB tubes, Lysis Buffer SF1, and FastPrep instruments.
  • Contaminants, including humic acids and RNA, are efficiently removed by treatment with Buffer SF2, while Buffer SF3 enables the specific binding of DNA to spin columns, without the need for RNase treatment.
  • The final DNA product is ready for downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome), without the need for further inhibitor removal steps.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type Up to 250 mg of Stool Samples
Includes 50 x Lysing Matrix YB, 45 mL Buffer SF1, 12 mL Buffer SF2, 35 mL Buffer SF3, 40 mL Buffer SF4, 40 mL Buffer SF5, 7.5 mL Buffer SF6, 50 x Column SF with collection tubes
Product Type DNA Extraction Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.